Assay Details
| Gene Symbol: psma6l | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_131795.2 | NP_571870.2 | 3 - 4 | 318 | 58 | |
| GenBank mRNA | AF354751.1 | - | 3 - 4 | 553 | 58 | |
| BC072719.1 | - | 3 - 4 | 318 | 58 | ||
| BC165117.1 | - | 3 - 4 | 284 | 58 | ||
More Information
Set Membership: |
|

psma6l
Genomic Map