Assay Details
| Gene Symbol: psma6l | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_131795.2 | NP_571870.2 | 4 - 5 | 479 | 96 | |
| GenBank mRNA | BC072719.1 | - | 4 - 5 | 479 | 96 | |
| BC165117.1 | - | 4 - 5 | 445 | 96 | ||
More Information
Set Membership: |
|

psma6l
Genomic Map