Assay Details
| Gene Symbol: emx1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_198144.2 | NP_937787.1 | 1 - 2 | 560 | 86 | |
| GenBank mRNA | AF534523.1 | - | 1 - 2 | 353 | 86 | |
| BC134029.1 | - | 1 - 2 | 560 | 86 | ||
More Information
Set Membership: |
|

emx1
Genomic Map