Assay Details
| Gene Symbol: ddx49 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_198365.1 | NP_938179.1 | 3 - 4 | 444 | 97 | |
| GenBank mRNA | BC047834.1 | - | 3 - 4 | 444 | 97 | |
| BC075762.1 | - | 3 - 4 | 418 | 97 | ||
More Information
Set Membership: |
|

ddx49
Genomic Map