Assay Details
| Gene Symbol: march5l | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_199739.3 | NP_956033.2 | 3 - 4 | 353 | 59 | |
| GenBank mRNA | BC066555.1 | - | 3 - 4 | 358 | 59 | |
More Information
Set Membership: |
|

march5l
Genomic Map