Assay Details
| Gene Symbol: march5l | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_199739.3 | NP_956033.2 | 5 - 6 | 669 | 111 | |
| GenBank mRNA | BC049066.1 | - | 4 - 5 | 458 | 111 | |
| BC066555.1 | - | 5 - 6 | 674 | 111 | ||
More Information
Set Membership: |
|

march5l
Genomic Map