Assay Details
| Gene Symbol: tnfaip1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_199799.1 | NP_956093.1 | 3 - 4 | 656 | 73 | |
| XM_005157506.4 | XP_005157563.1 | 3 - 4 | 610 | 73 | ||
| XM_017359218.2 | XP_017214707.1 | 3 - 4 | 546 | 73 | ||
| XM_021481342.1 | XP_021337017.1 | 4 - 5 | 624 | 73 | ||
| GenBank mRNA | BC044362.1 | - | 3 - 4 | 656 | 73 | |
| BC164349.1 | - | 2 - 3 | 439 | 73 | ||
More Information
Set Membership: |
|

tnfaip1
Genomic Map