Assay Details
| Gene Symbol: eif3s10 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_199820.2 | NP_956114.2 | 7 - 8 | 1210 | 63 | |
| XM_005156611.2 | XP_005156668.1 | 7 - 8 | 1121 | 63 | ||
| GenBank mRNA | BC059196.1 | - | 7 - 8 | 1222 | 63 | |
| BC066670.1 | - | 7 - 8 | 1210 | 63 | ||
More Information
Set Membership: |
|

eif3s10
Genomic Map