Assay Details
| Gene Symbol: paqr5a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_200187.1 | NP_956481.1 | 3 - 4 | 426 | 81 | |
| GenBank mRNA | BC045864.1 | - | 3 - 4 | 426 | 81 | |
More Information
Set Membership: |
|

paqr5a
Genomic Map