Assay Details
| Gene Symbol: paqr5a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_200187.1 | NP_956481.1 | 7 - 8 | 894 | 114 | |
| GenBank mRNA | BC045864.1 | - | 7 - 8 | 894 | 114 | |
More Information
Set Membership: |
|

paqr5a
Genomic Map