Assay Details
| Gene Symbol: diabloa | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_200346.1 | NP_956640.1 | 3 - 4 | 460 | 56 | |
| GenBank mRNA | BC053156.1 | - | 3 - 4 | 460 | 56 | |
More Information
Set Membership: |
|

diabloa
Genomic Map