Assay Details
| Gene Symbol: wipi1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_200391.1 | NP_956685.1 | 8 - 9 | 1034 | 82 | |
| XM_005164002.4 | XP_005164059.1 | 8 - 9 | 1100 | 82 | ||
| GenBank mRNA | BC053306.1 | - | 8 - 9 | 1034 | 82 | |
More Information
Set Membership: |
|

wipi1
Genomic Map