Assay Details
| Gene Symbol: xpa | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_200471.1 | NP_956765.1 | 3 - 4 | 379 | 74 | |
| XM_005159814.4 | XP_005159871.1 | 3 - 4 | 423 | 74 | ||
| XM_009306421.3 | XP_009304696.1 | 3 - 4 | 437 | 74 | ||
| XM_009306424.3 | XP_009304699.1 | 3 - 4 | 421 | 74 | ||
| GenBank mRNA | BC055179.1 | - | 3 - 4 | 379 | 74 | |
More Information
Set Membership: |
|

xpa
Genomic Map