Assay Details
| Gene Symbol: ndufa11 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_200525.2 | NP_956819.1 | 3 - 4 | 328 | 66 | |
| GenBank mRNA | BC055633.1 | - | 3 - 4 | 331 | 66 | |
More Information
Set Membership: |
|

ndufa11
Genomic Map