Assay Details
| Gene Symbol: unga | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_200974.1 | NP_957268.1 | 5 - 6 | 654 | 103 | |
| XM_017356048.2 | XP_017211537.1 | 4 - 5 | 761 | 103 | ||
| GenBank mRNA | BC045912.1 | - | 5 - 6 | 654 | 103 | |
| BC165165.1 | - | 5 - 6 | 588 | 103 | ||
More Information
Set Membership: |
|

unga
Genomic Map