Assay Details
| Gene Symbol: cnot4b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_201169.1 | NP_957463.1 | 6 - 7 | 938 | 114 | |
| GenBank mRNA | BC054913.1 | - | 6 - 7 | 938 | 114 | |
More Information
Set Membership: |
|

cnot4b
Genomic Map