Assay Details
| Gene Symbol: pou4f1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001312866.2 | NP_001299795.1 | 1 - 2 | 484 | 88 | |
| GenBank mRNA | AB188114.1 | - | 1 - 2 | 328 | 88 | |
| BC056326.1 | - | 1 - 2 | 463 | 88 | ||
| BC065954.1 | - | 1 - 2 | 322 | 88 | ||
| CK141066.1 | - | 1 - 2 | 505 | 88 | ||
| EH590787.1 | - | 1 - 2 | 334 | 88 | ||
More Information
Set Membership: |
|

pou4f1
Genomic Map