Assay Details
| Gene Symbol: pafah1b1a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_201345.1 | NP_958502.1 | 3 - 4 | 428 | 106 | |
| XM_005157575.4 | XP_005157632.1 | 3 - 4 | 421 | 106 | ||
| GenBank mRNA | BC053205.1 | - | 3 - 4 | 428 | 106 | |
| BC164856.1 | - | 2 - 3 | 150 | 106 | ||
| DQ141217.1 | - | 2 - 3 | 209 | 106 | ||
More Information
Set Membership: |
|

pafah1b1a
Genomic Map