Assay Details
| Gene Symbol: pdcl3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_205721.1 | NP_991284.1 | 581 | 68 | ||
| GenBank mRNA | AF175204.1 | - | 566 | 68 | ||
| BC064706.1 | - | 581 | 68 | |||
| BC152648.1 | - | 651 | 68 | |||
More Information
Set Membership: |
|

pdcl3
Genomic Map