Assay Details
| Gene Symbol: pard3ab | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_205735.1 | NP_991298.1 | 12 - 13 | 1895 | 67 | |
| GenBank mRNA | AF465629.1 | - | 12 - 13 | 1895 | 67 | |
| AY524776.2 | - | 12 - 13 | 1926 | 67 | ||
| AY524777.2 | - | 12 - 13 | 1926 | 67 | ||
| BC162460.1 | - | 12 - 13 | 1745 | 67 | ||
| BC162465.1 | - | 12 - 13 | 1745 | 67 | ||
More Information
Set Membership: |
|

pard3ab
Genomic Map