Assay Details
| Gene Symbol: ptk2bb | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_212570.1 | NP_997735.1 | 14 - 15 | 1419 | 72 | |
| GenBank mRNA | AJ490510.2 | - | 14 - 15 | 1419 | 72 | |
More Information
Set Membership: |
|

ptk2bb
Genomic Map