Assay Details
| Gene Symbol: ptk2bb | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_212570.1 | NP_997735.1 | 16 - 17 | 1606 | 84 | |
| GenBank mRNA | AF400274.1 | - | 2 - 3 | 203 | 84 | |
| AJ490510.2 | - | 16 - 17 | 1606 | 84 | ||
More Information
Set Membership: |
|

ptk2bb
Genomic Map