Assay Details
| Gene Symbol: ptk2bb | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_212570.1 | NP_997735.1 | 1 - 2 | 290 | 92 | |
| GenBank mRNA | AJ490510.2 | - | 1 - 2 | 290 | 92 | |
| BC134027.1 | - | 2 - 3 | 321 | 92 | ||
More Information
Set Membership: |
|

ptk2bb
Genomic Map