Assay Details
| Gene Symbol: ptk2bb | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_212570.1 | NP_997735.1 | 6 - 7 | 762 | 77 | |
| GenBank mRNA | AJ490510.2 | - | 6 - 7 | 762 | 77 | |
| BC134027.1 | - | 7 - 8 | 793 | 77 | ||
More Information
Set Membership: |
|

ptk2bb
Genomic Map