Assay Details
| Gene Symbol: rab20 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_212771.1 | NP_997936.1 | 1 - 2 | 412 | 139 | |
| GenBank mRNA | BC053222.1 | - | 1 - 2 | 412 | 139 | |
| BC165051.1 | - | 1 - 2 | 173 | 139 | ||
More Information
Set Membership: |
|

rab20
Genomic Map