Assay Details
| Gene Symbol: mao | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_212827.3 | NP_997992.2 | 7 - 8 | 834 | 98 | |
| GenBank mRNA | AY185211.3 | - | 7 - 8 | 778 | 98 | |
| BC070013.1 | - | 7 - 8 | 834 | 98 | ||
More Information
Set Membership: |
|

mao
Genomic Map