Assay Details
| Gene Symbol: nlk1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_212956.1 | NP_998121.1 | 2 - 3 | 514 | 79 | |
| GenBank mRNA | AY562552.1 | - | 2 - 3 | 514 | 79 | |
| BC093121.1 | - | 2 - 3 | 79 | |||
| BC164359.1 | - | 1 - 2 | 79 | |||
More Information
Set Membership: |
|

nlk1
Genomic Map