Assay Details
| Gene Symbol: nlk1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_212956.1 | NP_998121.1 | 7 - 8 | 1099 | 71 | |
| GenBank mRNA | AY562552.1 | - | 7 - 8 | 1099 | 71 | |
More Information
Set Membership: |
|

nlk1
Genomic Map