Assay Details
| Gene Symbol: nlk1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_212956.1 | NP_998121.1 | 9 - 10 | 1292 | 59 | |
| GenBank mRNA | AY562552.1 | - | 9 - 10 | 1292 | 59 | |
More Information
Set Membership: |
|

nlk1
Genomic Map