Assay Details
| Gene Symbol: nrp2b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_212966.1 | NP_998131.1 | 16 - 17 | 3044 | 78 | |
| GenBank mRNA | AY351913.1 | - | 16 - 17 | 2541 | 78 | |
| AY428802.1 | - | 16 - 17 | 3044 | 78 | ||
| AY437404.1 | - | 16 - 17 | 3004 | 78 | ||
| BC163589.1 | - | 16 - 17 | 2859 | 78 | ||
| KM655620.1 | - | 16 - 17 | 2454 | 78 | ||
More Information
Set Membership: |
|

nrp2b
Genomic Map