Assay Details
| Gene Symbol: nrp2b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_212966.1 | NP_998131.1 | 8 - 9 | 1874 | 65 | |
| XM_017357663.2 | XP_017213152.1 | 8 - 9 | 2003 | 65 | ||
| GenBank mRNA | AY351913.1 | - | 8 - 9 | 1371 | 65 | |
| AY428802.1 | - | 8 - 9 | 1874 | 65 | ||
| AY437404.1 | - | 8 - 9 | 1834 | 65 | ||
| BC163576.1 | - | 8 - 9 | 1689 | 65 | ||
| BC163589.1 | - | 8 - 9 | 1689 | 65 | ||
| KM655620.1 | - | 8 - 9 | 1284 | 65 | ||
More Information
Set Membership: |
|

nrp2b
Genomic Map