Assay Details
| Gene Symbol: gdf11 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_212975.1 | NP_998140.1 | 1 - 2 | 688 | 68 | |
| XM_005155799.4 | XP_005155856.1 | 1 - 2 | 726 | 68 | ||
| GenBank mRNA | AF411599.3 | - | 1 - 2 | 695 | 68 | |
| BC134028.1 | - | 1 - 2 | 688 | 68 | ||
| KM655715.1 | - | 1 - 2 | 418 | 68 | ||
More Information
Set Membership: |
|

gdf11
Genomic Map