Assay Details
| Gene Symbol: mfsd14a1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_213527.2 | NP_998692.2 | 3 - 4 | 422 | 70 | |
| GenBank mRNA | BC045310.1 | - | 3 - 4 | 420 | 70 | |
| BC097075.1 | - | 3 - 4 | 416 | 70 | ||
More Information
Set Membership: |
|

mfsd14a1
Genomic Map