Assay Details
| Gene Symbol: rab11fip4a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001002533.2 | NP_001002533.2 | 13 - 14 | 1703 | 98 | |
| XM_009299531.3 | XP_009297806.1 | 13 - 14 | 1701 | 98 | ||
| XM_009299532.3 | XP_009297807.1 | 11 - 12 | 1611 | 98 | ||
| GenBank mRNA | AB208638.1 | - | 13 - 14 | 1703 | 98 | |
| AB208639.1 | - | 11 - 12 | 1575 | 98 | ||
More Information
Set Membership: |
|

rab11fip4a
Genomic Map