Assay Details
| Gene Symbol: rab11fip4a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001002533.2 | NP_001002533.2 | 7 - 8 | 984 | 81 | |
| XM_009299531.3 | XP_009297806.1 | 7 - 8 | 982 | 81 | ||
| XM_009299532.3 | XP_009297807.1 | 5 - 6 | 892 | 81 | ||
| GenBank mRNA | AB208638.1 | - | 7 - 8 | 984 | 81 | |
| AB208639.1 | - | 5 - 6 | 856 | 81 | ||
More Information
Set Membership: |
|

rab11fip4a
Genomic Map