Assay Details
| Gene Symbol: rab11fip4a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001002533.2 | NP_001002533.2 | 8 - 9 | 1084 | 70 | |
| XM_009299531.3 | XP_009297806.1 | 8 - 9 | 1082 | 70 | ||
| XM_009299532.3 | XP_009297807.1 | 6 - 7 | 992 | 70 | ||
| GenBank mRNA | AB208638.1 | - | 8 - 9 | 1084 | 70 | |
| AB208639.1 | - | 6 - 7 | 956 | 70 | ||
More Information
Set Membership: |
|

rab11fip4a
Genomic Map