Assay Details
| Gene Symbol: pbldp | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001003759.1 | NP_001003759.1 | 5 - 6 | 492 | 70 | |
| GenBank mRNA | BC079517.1 | - | 5 - 6 | 492 | 70 | |
More Information
Set Membership: |
|

pbldp
Genomic Map