Assay Details
| Gene Symbol: lhx6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001004015.2 | NP_001004015.1 | 8 - 9 | 1351 | 64 | |
| XM_009305145.3 | XP_009303420.1 | 8 - 9 | 2967 | 64 | ||
| GenBank mRNA | AY664403.1 | - | 8 - 9 | 1352 | 64 | |
| BC162225.1 | - | 8 - 9 | 1292 | 64 | ||
More Information
Set Membership: |
|

lhx6
Genomic Map