Assay Details
| Gene Symbol: ap1s3b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001004635.1 | NP_001004635.1 | 3 - 4 | 395 | 110 | |
| GenBank mRNA | BC081385.1 | - | 3 - 4 | 395 | 110 | |
| BC150444.1 | - | 3 - 4 | 110 | |||
More Information
Set Membership: |
|

ap1s3b
Genomic Map