Assay Details
| Gene Symbol: tmem248 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001013530.1 | NP_001013548.1 | 5 - 6 | 1014 | 63 | |
| XM_005168624.4 | XP_005168681.1 | 5 - 6 | 1019 | 63 | ||
| GenBank mRNA | BC090900.1 | - | 5 - 6 | 1014 | 63 | |
More Information
Set Membership: |
|

tmem248
Genomic Map