Assay Details
| Gene Symbol: afmid | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001017677.1 | NP_001017677.1 | 9 - 10 | 835 | 107 | |
| XM_009306326.3 | XP_009304601.1 | 10 - 11 | 1097 | 107 | ||
| GenBank mRNA | BC093330.1 | - | 9 - 10 | 835 | 107 | |
More Information
Set Membership: |
|

afmid
Genomic Map