Assay Details
| Gene Symbol: bbox1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001017717.1 | NP_001017717.1 | 4 - 5 | 837 | 75 | |
| XM_005168903.4 | XP_005168960.1 | 4 - 5 | 842 | 75 | ||
| XM_005168904.4 | XP_005168961.1 | 4 - 5 | 836 | 75 | ||
| XM_021477587.1 | XP_021333262.1 | 5 - 6 | 938 | 75 | ||
| GenBank mRNA | BC093246.1 | - | 4 - 5 | 837 | 75 | |
More Information
Set Membership: |
|

bbox1
Genomic Map