Assay Details
| Gene Symbol: mpeg1.2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001020586.2 | NP_001018422.2 | 1 - 2 | 506 | 66 | |
| GenBank mRNA | BC095229.1 | - | 1 - 2 | 507 | 66 | |
More Information
Set Membership: |
|

mpeg1.2
Genomic Map