Assay Details
| Gene Symbol: c6ast4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001025180.1 | NP_001020351.1 | 8 - 9 | 833 | 81 | |
| XM_009303326.3 | XP_009301601.1 | 9 - 10 | 873 | 81 | ||
| GenBank mRNA | BC097232.1 | - | 8 - 9 | 833 | 81 | |
More Information
Set Membership: |
|

c6ast4
Genomic Map