Assay Details
| Gene Symbol: rdh5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001030101.1 | NP_001025272.1 | 4 - 5 | 760 | 115 | |
| XM_005173961.4 | XP_005174018.1 | 4 - 5 | 815 | 115 | ||
| XM_009296105.3 | XP_009294380.1 | 4 - 5 | 821 | 115 | ||
| GenBank mRNA | BC097151.1 | - | 4 - 5 | 760 | 115 | |
| DQ000308.1 | - | 3 - 4 | 603 | 115 | ||
More Information
Set Membership: |
|

rdh5
Genomic Map