Assay Details
| Gene Symbol: rdh5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001030101.1 | NP_001025272.1 | 5 - 6 | 910 | 85 | |
| XM_005173961.4 | XP_005174018.1 | 5 - 6 | 965 | 85 | ||
| XM_009296105.3 | XP_009294380.1 | 5 - 6 | 971 | 85 | ||
| GenBank mRNA | BC097151.1 | - | 5 - 6 | 910 | 85 | |
More Information
Set Membership: |
|

rdh5
Genomic Map