Assay Details
| Gene Symbol: rhoj | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001044801.2 | NP_001038266.1 | 4 - 5 | 538 | 63 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

rhoj| Gene Symbol: rhoj | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001044801.2 | NP_001038266.1 | 4 - 5 | 538 | 63 | |
Set Membership: |
|