Assay Details
| Gene Symbol: si:ch211-106n13.3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001045014.1 | NP_001038479.1 | 1 - 2 | 580 | 55 | |
| XM_021480844.1 | XP_021336519.1 | 1 - 2 | 580 | 55 | ||
More Information
Set Membership: |
|

si:ch211-106n13.3
Genomic Map