Assay Details
| Gene Symbol: si:ch211-106n13.3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001045014.1 | NP_001038479.1 | 21 - 22 | 2155 | 86 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

si:ch211-106n13.3| Gene Symbol: si:ch211-106n13.3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001045014.1 | NP_001038479.1 | 21 - 22 | 2155 | 86 | |
Set Membership: |
|