Assay Details
| Gene Symbol: dgkaa | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001045315.1 | NP_001038780.1 | 16 - 17 | 1541 | 111 | |
| XM_005162328.4 | XP_005162385.1 | 17 - 18 | 1695 | 111 | ||
| GenBank mRNA | BC098607.1 | - | 16 - 17 | 1537 | 111 | |
| BC117645.1 | - | 16 - 17 | 1541 | 111 | ||
More Information
Set Membership: |
|

dgkaa
Genomic Map